Learn More
Gibco™ B-27™ Supplement (50X), minus vitamin A
Suitable for use in most neuronal cell applications
Supplier: Gibco™ 12587001
Description
A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Vitamin A is a customized version of the original B-27 Supplement without vitamin A (retinyl acetate). B-27 Supplement Minus Vitamin A is a serum-free supplement ideal for the cultivation of neural progenitor and stem cells, either as neurospheres in suspension or in adherent monolayer culture, without inducing differentiation. B-27 Supplement Minus Vitamin A is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.
B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media
Specifications
Store in freezer (-5 to -20°C) and protect from light. | |
Serum-free | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Sterile | |
100 mL | |
Serum-Free |
Neural Progenitor Cells | |
50 X | |
Liquid | |
Serum-Free | |
B-27™ | |
Dry Ice | |
Endotoxin, Performance, Sterility |
For Research Use Only. Not for use in diagnostic procedures.