missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ B-7569 Vinyl GHS L4A Diamond Labels for BBP85 Printer

Allows printer to create 2-color GHS/CLP chemical labels in one print pass

$214.10 - $320.44

Specifications

Material Vinyl (B-7569)
Product Type GHS Diamond Label
Display Text L4A Diamond + Text
For Use With (Equipment) BBP™85 Printer
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
19-000-570
View Documents Promotion Details
Brady™
130776
White, Red
Each for $320.44
 
19-000-122
View Documents Promotion Details
Brady™
130770
Red, Yellow, Blue, White
Each for $214.10
 
Description

Description

  • White labels with pre-printed diamonds
  • Indoor/outdoor grade withstands exposure to moisture, chemicals and handling
  • Available in L1, L2, L3A, L3B, L4A and L4B diamond legends

Specifications

Specifications

Vinyl (B-7569)
GHS Diamond Label
L4A Diamond + Text
BBP™85 Printer
SDS
missing translation for 'documents'

missing translation for 'documents'