Büchner Funnels
$77.46 - $77.46
Specifications
Material | Porcelain |
---|---|
For Use With (Application) | Designed to withstand sudden temperature changes of 1050°C without cracks, explosion or deformation |
Includes | Perforated filter |
Product Type | Buchner Funnel |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
S34596-1
|
Customer Channels Group (Ccg)
C146 |
4 Boxes |
Pack of 4 Each for $77.46
Case of 12 Pack for $929.52
|
|
|||||
Description
Fired twice for added strength and durability
- Easy to clean
- Standard size perforated area fits most filter paper
Specifications
Porcelain | |
Designed to withstand sudden temperature changes of 1050°C without cracks, explosion or deformation | |
Perforated filter | |
Buchner Funnel |