missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ beta-Glucosidase Assay Kit
Colorimetric kinetic determination of beta-glucosidase activity
Supplier: Abnova™ KA1611
Comprend:
- Assay buffer: (pH 7.0), 24mL
- β-NPG substrate, 1mL
- Calibrator: (equivalent to 250U/L), 10mL
Spécifications
| Store the kit at -20°C. | |
| Colorimetric | |
| 24mL assay buffer (pH 7), 1mL beta-NPG substrate, 10mL calibrator (equivalent to 250U/L) | |
| 20 μL |
| beta-Glucosidase Assay Kit is a colorimetric kinetic determination of beta-Glucosidase activity. | |
| Functional Study | |
| 1 Kit | |
| Activity Detection Assay Kit |