missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Motic™ BA310E Compound LED Microscopes

Committed to the concept of continuously improving its models, Motic™â€™s Elite series microscopes feature the Infinity Optical System and improved CCIS™ EC Plan Objective Lenses. Provides superior image clarity and contrast.

$4519.06 - $5039.74

Specifications

Voltage 6 V
Magnification Power 10X
Objective Types CCIS™ EC Plan Achromat Objectives EC PL (4X, 10X, 40X(S), 100X(S-oil))
Nosepiece Reversed Quadruple
Lighting Type LED
View More Specs

Products 2
Catalog Number Mfr. No. Head Style Price Quantity  
Catalog Number Mfr. No. Head Style Price Quantity  
01-258-023
View Documents
Motic™
1100100402631
Binocular
Each for $4,519.06
 
01-258-024
View Documents
Motic™
1100100402641
Trinocular
Each for $5,039.74
 
Description

Description

These models introduce a new rackless stage concept without a prominent gear rack to allow for a more convenient use of the X/Y movement.

  • Allow easy interchangeability of its halogen bulb with LED modules
  • Swiveling, 30° inclined Siedentopf type
  • Reversed quintuple revolving nosepiece
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built in low position coaxial rackless stage (Right-hand control)
  • Diopter Adjustment: ±5
Specifications

Specifications

6 V
CCIS™ EC Plan Achromat Objectives EC PL (4X, 10X, 40X(S), 100X(S-oil))
LED
10X
Reversed Quadruple
WF 10X/20 mm
SDS
missing translation for 'documents'

missing translation for 'documents'