Mopec Back Draft Countertop Dissection Work Bench Grossing Station
Available in four highly customizable models, these back draft vented workstations help control odors, gases and formalin fumes. Mopec™ Back Draft Countertop Dissection Work Bench Grossing Station ensures a safe laboratory working environment.
Supplier:  Mopec HL096
 View more versions of this product
Catalog No. 22444134
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Countertop constructed of 14 gauge stainless steel with a recessed edge
  • Includes vent tie-in
  • Adjustable leveling feet
  • GFCI outlet
Warranty
1 Year Warranty
Specifications
Specifications
Ventilated Grossing Station
30 in.
62 in.
157.48 cm
Stainless Steel (14 ga.)
76.2 cm
96 in.
243.84 cm