missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ BactiSwab™: Liquid Stuart Plastic Shaft
Thermo Scientific™ Remel BactiSwab™ Liquid Stuart, Plastic Shaft is supplied with a single or double plastic shaft.
$286.86 - $286.86
Specifications
Format | Tube |
---|---|
Media Type | Liquid Stuart |
Description | Liquid Stuarts |
For Use With (Application) | Healthcare |
Shaft Material | Plastic |
Catalog Number | Mfr. No. | SwabConfiguration | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | SwabConfiguration | Price | Quantity | ||||||
R723115
|
Thermo Scientific™
RE723115 |
Single Applicator |
Pack of 50 Each for $286.86
|
|
||||||
Description
Thermo Scientific™ Remel BactiSwab™ Liquid Stuart, Plastic Shaft is supplied with a single or double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.Specifications
Tube | |
Liquid Stuarts | |
Plastic |
Liquid Stuart | |
Healthcare |