missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Malt Extract

The water-soluble portion of malted barley.

$124.04 - $2160.75

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Line Bacto
Source Animal origin-free, malted barley
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
218610TF
View Documents
Gibco™
218610
10 kg
Each for $2,160.75
Only null left
 
DF0186-17-7
View Documents
Gibco™
218630
500 g
Each for $124.04
Only null left
 
Description

Description

  • Bacto Malt Extract is the water-soluble portion of malted barley
  • Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium
  • It has been used successfully in microbial fermentation applications
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Bacto
Animal origin-free, malted barley
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'