missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Bacto™ Yeast Extract, technical
Suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications.
$183.29 - $3260.05
Specifications
Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Culture Type | Mammalian, Insect, Microbial |
Product Type | Bacterial Media |
For Use With (Application) | Dehydrated culture media |
Product Line | Bacto™ |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
DF0886-17-0
|
Gibco™
288620 |
500 g |
Each for $183.29
|
|
|||||
DF0886-08-1
|
Gibco™
288610 |
10 kg |
Each for $3,260.05
|
|
|||||
Description
- Concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
- Bacto Yeast Extract is an important ingredient for the microbiological assay of vitamins
- B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
- Developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Bacterial Media | |
Bacto™ |
Mammalian, Insect, Microbial | |
Dehydrated culture media | |
Animal origin-free |