Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
$19.82 - $109.53
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Includes | Price | Quantity | ||||
05-885B
|
Bal Supply
20112WOS |
For Ball to Socket Joint Size: 12/1 and 12/5 | 12/Pk. | Without the screw device |
Each for $19.82
Case of 12 Each for $237.84
|
|
||||
05-885D
|
Bal Supply
20118WOS |
For 18/7 to 18/9 Joints | 12/Pk. | Without screw locking device |
Each for $21.56
Case of 12 Each for $258.72
|
|
||||
05-885F
|
Bal Supply
20128WS |
For 28/11 to 28/15 Joints | 12/Pk. | Screw locking device |
Each for $29.12
Case of 12 Each for $349.44
|
|
||||
05-885C
|
Bal Supply
20112WS |
For Ball to Socket Joint Size: 12/1 and 12/5 | 12/Pk. | Screw locking device |
Each for $27.35
Case of 12 Each for $328.20
|
|
||||
05-885E
|
Bal Supply
20118WS |
For 18/7 to 18/9 Joints | 12/Pk. | Screw locking device |
Each for $28.18
Case of 12 Each for $338.16
|
|
||||
05-885G
|
Bal Supply
20135WS |
For 35/20 to 35/25 Joints | 12/Pk. | Screw locking device |
Each for $38.91
Case of 12 Each for $466.92
|
|
||||
05-885K
|
Bal Supply
20165WS |
For 65/40 Joints | 6/Pk. | Screw locking device |
Each for $109.53
Case of 6 Each for $657.18
|
|
||||
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
- 05-885C and 05-885E through K have screw-locking device
- Work well on both O-ring and ball-and-socket joints
- Others are intended for ball-and-socket joints only