Eisco™ Ballistic Pendulum
The Ballistic Pendulum contains everything needed to measure the momentum of a projected object.
Supplier: Eisco™ BLPN14
Description
This unit can be used to aid in the understanding of momentum, velocity, conservation of momentum and energy, potential energy, and kinetic energy. Its sturdy construction and clear labels make it easy to use.- Includes ballistic launcher, rigid arm pendulum, angle backdrop, steel ball, and release mechanism
- Convenient angle backdrop makes measurements easier
- Black in color with white labeling
- Rigid arm pendulum with rubber head
- All pieces are connected and can be moved as one unit
Specifications
Pendulum, Ballistic | |
Ballistic launcher, rigid arm pendulum, angle back drop, steel ball, release mechanism |