Aspen Surgical™ Bard-Parker™ Protected Disposable Scalpel
Features one-handed, locking retractable shield
Supplier:  Aspen Surgical™ 372611
 View more versions of this product
Catalog No. 02-688-79
Explore available promotions
Description
Description
Features one-handed, locking retractable shield
  • Designed to minimize the risk of surgical blade injuries
  • Features a BD Bard-Parker stainless-steel blade, nonslip grip, and a centimeter scale on the plastic, surgical-green handle
  • Clear protective shield locks both when the blade is covered for protection before and after use, and when blade is uncovered for security and control during use
  • Top button provides tactile sensation as well as an audible click to confirm lock; allows for left or right hand activation
  • Indicator line offers visual confirmation of lock
  • Autoclavable for procedural kits and on-site sterilization
Specifications
Specifications
Stainless Steel
Tissue Separation and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Single-use
OSHA
Safety Scalpel
# 11
Sterile
Autoclavable