Learn More
Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 317861000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
99+% | |
7440-39-3 | |
99.0 | |
99+% | |
Ba | |
100 g | |
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom | |
[Ba] | |
137.33 | |
CHEBI:32595 | |
≥99% | |
Rod |
Barium | |
725.0°C | |
1640.0°C | |
100.0 | |
Ba | |
MFCD00134031 | |
15, 960 | |
DSAJWYNOEDNPEQ-UHFFFAOYSA-N | |
barium | |
5355457 | |
137.34 | |
Plastic bottle |
Chemical Identifiers
7440-39-3 | |
137.33 | |
DSAJWYNOEDNPEQ-UHFFFAOYSA-N | |
5355457 | |
barium |
Ba | |
MFCD00134031 | |
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom | |
CHEBI:32595 | |
[Ba] |
Safety and Handling
GHS H Statement:
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
In contact with water releases flammable gas.
Reacts violently with water.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only