Learn More
Barium acetate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01112614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium acetate | |
White | |
25 g | |
C4H6BaO4 | |
MFCD00012447 | |
3693411 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
[Ba++].CC([O-])=O.CC([O-])=O | |
255.42 | |
255.43 | |
Puratronic™ | |
2.47 g/mL |
543-80-6 | |
Crystalline | |
(metals basis) | |
Ba(OOCCH3)2 | |
UN1564 | |
14,965 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
barium(2+) diacetate | |
10980 | |
99.999% | |
Odorless |
Chemical Identifiers
543-80-6 | |
255.42 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
10980 |
C4H6BaO4 | |
MFCD00012447 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
[Ba++].CC([O-])=O.CC([O-])=O |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 208-849-0
RTECSNumber : AF4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only