missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Barium Chloride Anhydrous (Laboratory), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLB31500
Specifications
1560°C | |
960°C | |
White | |
500 g | |
BaCl2 | |
UN 1564 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
barium(2+) dichloride | |
25204 | |
208.23 | |
Laboratory | |
Poly Bottle |
Barium Chloride Anhydrous | |
10361-37-2 | |
Solid | |
≥97% | |
MFCD00003445 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
[Cl-].[Cl-].[Ba++] | |
208.23 | |
CHEBI:63317 | |
≥97% | |
Pass Test | |
3.86g/cm³ |
Chemical Identifiers
10361-37-2 | |
208.23 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
25204 | |
barium(2+) dichloride |
BaCl2 | |
MFCD00003445 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
CHEBI:63317 | |
[Cl-].[Cl-].[Ba++] |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison