Learn More
Barium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423450025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1560.0°C | |
962.0°C | |
White | |
Crystals | |
99% min. (Complexometry) | |
BaCl2·2H2O | |
15, 967 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,practically insoluble in ethanol,acetone and,ethyl acetate | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
≥99% | |
14.0% to 16.0% (150°C) |
Barium chloride dihydrate | |
10326-27-9 | |
5.2 to 8.2 (5% soln. at 25°C) | |
2.5 kg | |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.28 | |
ACS Reagent | |
Plastic bottle |
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
barium(2+) dihydrate dichloride |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 | |
O.O.[Cl-].[Cl-].[Ba++] |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per
GHS Signal Word: Danger
RTECSNumber : CQ8751000
RUO – Research Use Only