missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Barium crystalline dendritic solid, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01445414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
7440-39-3 | |
99.9% (Metals basis) | |
MFCD00134031 | |
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom | |
[Ba] | |
137.33 | |
CHEBI:32595 | |
Crystalline Dendritic Solid |
Barium | |
Silver Gray solid | |
Ba | |
25 g | |
DSAJWYNOEDNPEQ-UHFFFAOYSA-N | |
barium | |
5355457 | |
Odorless |
Chemical Identifiers
7440-39-3 | |
137.33 | |
DSAJWYNOEDNPEQ-UHFFFAOYSA-N | |
5355457 | |
barium |
Ba | |
MFCD00134031 | |
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom | |
CHEBI:32595 | |
[Ba] |
Safety and Handling
P223-P231+P232-P280-P301+P310-P331-P335+P334-P370+P378q-P501c
H260-H304
DOTInformation : DOT Class: 4.3, Packaging Group: II
EINECSNumber : 231-149-1
RTECSNumber : CQ8370000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only