Learn More
Barium hydroxide octahydrate, 97%, Thermo Scientific Chemicals
Alfa Aesar Barium hydroxide octahydrate, 97%, Quantity: 500g, CAS: 12230-71-6, Melting Point: 78 deg.C, Molecular Formula: BaH18O10, Molecular Weight: 315.461, Percent Purity: 97%
$84.81 - $950.84
Chemical Identifiers
CAS | 12230-71-6 |
---|---|
Molecular Formula | BaH18O10 |
Molecular Weight (g/mol) | 315.46 |
MDL Number | MFCD00149152 |
InChI Key | ZUDYPQRUOYEARG-UHFFFAOYSA-L |
Synonym | barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate |
PubChem CID | 17749109 |
SMILES | O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1271436
|
Thermo Scientific Chemicals
A1271436 |
500 g |
Each for $84.81
|
|
|||||
AAA127140E
|
Thermo Scientific Chemicals
A127140E |
2500 g |
Each for $304.18
|
|
|||||
AAA127140C
|
Thermo Scientific Chemicals
A127140C |
10000 g |
Each for $950.84
|
|
|||||
Description
Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBarium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.
Solubility
Soluble in water (72 mg/ml at 20°C), methanol, ethanol (slightly), and water (56 mg/ml at 15°C). Insoluble in acetone (practically)
Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
Chemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
Specifications
78°C | |
500 g | |
Ba(OH)2·8H2O | |
UN3262 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+);dihydroxide;octahydrate | |
17749109 | |
97% | |
Air Sensitive | |
Barium hydroxide octahydrate |
12230-71-6 | |
BaH18O10 | |
MFCD00149152 | |
14,977 | |
Soluble in water (72mg/ml at 20°C),methanol,ethanol (slightly),and water (56mg/ml at 15°C). Insoluble in acetone (practically) | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.48 (171.36 Anhydrous) | |
Odorless | |
2.18 |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 241-234-5
RTECSNumber : CQ9200000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only