missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Barium hydroxide solution, Volumetric, 0.05Â M Ba(OH)2 (0.1N), Honeywell Fluka™
Volumetric, 0.05Â M Ba(OH)2 (0.1N)
$211.05 - $211.05
Chemical Identifiers
CAS | 17194-00-2 |
---|---|
Molecular Formula | BaH2O2 |
Molecular Weight (g/mol) | 171.341 |
InChI Key | RQPZNWPYLFFXCP-UHFFFAOYSA-L |
Synonym | barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous |
PubChem CID | 6093286 |
ChEBI | CHEBI:32592 |
IUPAC Name | barium(2+);dihydroxide |
SMILES | [OH-].[OH-].[Ba+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6002589
|
Honeywell-Fluka
350501L |
1 L |
Each for $211.05
|
|
|||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
17194-00-2 | |
171.341 | |
barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous | |
CHEBI:32592 | |
[OH-].[OH-].[Ba+2] |
BaH2O2 | |
RQPZNWPYLFFXCP-UHFFFAOYSA-L | |
6093286 | |
barium(2+);dihydroxide |
Specifications
17194-00-2 | |
BaH2O2 | |
MFCD00003443 | |
barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous | |
[OH-].[OH-].[Ba+2] | |
171.341 | |
CHEBI:32592 | |
Barium hydroxide solution |
1 L | |
Ba(OH)2 | |
NONH for all modes of transport | |
RQPZNWPYLFFXCP-UHFFFAOYSA-L | |
barium(2+);dihydroxide | |
6093286 | |
171.34g/mol |