Learn More
Barium perchlorate trihydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011656A1
Description
Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium perchlorate trihydrate | |
Crystalline | |
BaCl2H6O11 | |
UN1447 | |
barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate | |
NBHDMFAHVJWOHD-UHFFFAOYSA-L | |
barium(2+);diperchlorate;trihydrate | |
53249218 | |
Reagent | |
1.533 |
10294-39-0 | |
1 kg | |
MFCD00149148 | |
14,987 | |
Soluble in water, methanol; Slightly soluble in ethanol, ethyl acetate, acetone; Insoluble in ether | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] | |
390.264 | |
390.29 (336.24 Anhydrous) | |
Odorless | |
2.74 g/mL |
Chemical Identifiers
10294-39-0 | |
390.264 | |
NBHDMFAHVJWOHD-UHFFFAOYSA-L | |
53249218 | |
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
BaCl2H6O11 | |
MFCD00149148 | |
barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate | |
barium(2+);diperchlorate;trihydrate |
Safety and Handling
GHS H Statement
H271-H302-H332
May cause fire or explosion
strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM PERCHLORATE, SOLID
EINECSNumber : 236-710-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only