Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Supplier: Thermo Scientific™ D50281
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
Deionized water feed ; Ultralow organics, type I water | |
Nanopure Water Purifications System | |
Deionized Water Feed |