missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix
Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.
Supplier: Corning™ 16405CV
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
- Serum-free growth may be possible with appropriate protein supplements
- Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
- Formulation of DMEM/Ham's F-12 contains L-glutamine
- Sterile
Specifications
2° to 8°C | |
Liquid | |
Sterile | |
No Phenol Red | |
To assess growth promotion and cytotoxicity characteristics | |
6 x 500 mL |
1 X | |
DMEM and Ham's F-12, 50/50 Mix | |
Glutamine | |
<0.25EU/mL | |
7 to 7.4 |
IVD Product