Basix™ Gel-Loading Tips
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
 View more versions of this product
Catalog No. 13-611-102
Explore available promotions
Description
Description
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everdayliquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
    Round gel-loading tipEasily load gels 0.6mmApplications include Agarose DNA gels, 2-D and SDS PAGE, capillary gels and sequencingCertified by independent laboratory
Specifications
Specifications
10 μL
Natural
5.7cm
Rack
Basix
Non-sterile
2.25 in.
DNase, RNase, and DNA Free
Gel Loading, Round
Polypropylene