missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Basix™ Polypropylene Conical Centrifuge Tubes
Sterile tubes with graduated divisions
Description
- Made from polypropylene
- RCF rating of 12,000xG
- Available in rack or sleeve
Specifications
DNase/RNase free | |
Polypropylene | |
Sterile | |
Conical Centrifuge Tube |
50 mL | |
Rack | |
2.5 mL |