missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ CultureSwab EZ™ and CultureSwab EZ II™
A patented media-free collection system designed to collect aerobic specimens and maintain viability during transport
$310.50 - $578.11
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | For collecting and transporting aerobic microbiological specimens |
Quantity | 100/Pk. |
Shaft Material | Plastic |
Storage Requirements | 15°C to 30°C |
Catalog Number | Mfr. No. | Includes | Product Type | SwabConfiguration | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Product Type | SwabConfiguration | Price | Quantity | |||
B220144
|
BD
220144 |
Polyurethane single sponge swab, Plastic shaft, cap, inserted into Plastic transport tube | Culture Swab™ EZ System | Single Applicator |
Pack of 100 Each for $310.50
|
|
|||
B220145
|
BD
220145 |
Polyurethane dual sponge swab, Plastic shaft, cap, inserted into Plastic transport tube | Culture Swab™ EZ II System | Dual Applicator |
Pack of 100 Each for $578.11
|
|
|||
Description
- No specimen dilution with this dry system and no ampule to crush
- Transport system eliminates the organism overgrowth and nonviables typically present in agar-based systems
- Aerobic microorganisms remain viable for approximately 48 hours
- Features rounded tube bottom for easier handling, and water-resistant film packing with tamper-evident seal
- BD CultureSwab EZ II is similar to BD CultureSwab EZ except formatted as a dual swab system, allowing for simultaneous gram staining and other direct antigen testing
Specifications
Sterile | |
100/Pk. | |
15°C to 30°C |
For collecting and transporting aerobic microbiological specimens | |
Plastic |