BD BBL™ Gram Stain Kits
Used to stain microorganisms from cultures or specimens by the differential Gram method
$343.45 - $343.45
Specifications
Quantity 1 Kit
Certifications/Compliance CE
Includes 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Stain Type Gram Stain
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
B4312539
View Documents
BD
212539
Gram Stain Kit (Stabilized)
Each for $343.45
Description
Description
  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life
Specifications
Specifications
1 Kit
250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
Gram Stain
CE
Used to stain microorganisms from cultures or specimens by the differential gram method
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'