missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ MycoPrep™ Mycobacterial System Digestion/Decontamination Kits
No weighing and no messy mixing
$399.16 - $748.68
Specifications
Product Type | Specimen Collection Kit |
---|---|
Certifications/Compliance | CE |
For Use With (Application) | For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis |
Storage Requirements | 15°C to 25°C |
Catalog Number | Mfr. No. | Volume (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Includes | Price | Quantity | ||||
B4340862
|
BD
240862 |
75 mL | 10 x 75 mL bottles of NALC-NaOH solution and 5 packets Phosphate Buffer (ph 6.8) |
Each for $399.16
|
|
||||
B4340863
|
BD
240863 |
150 mL | 10 x 150 mL bottles of NALC-NaOH solution and 5 packets Phosphate Buffer (ph 6.8) |
Each for $748.68
|
|
||||
Description
- Provide one of the most widely used, CDC-recommended formulations for effective mycobacterial specimen processing
- Convenient, all-inclusive kits are an alternative to traditional methods of preparing specimen digestion/decontamination reagents
- Complete with bottles of NALC-NaOH Solution and pre-measured packets of Phosphate Buffer (pH 6.8)
- Ensure standardization by eliminating the need to weigh, reconstitute or inventory separate ingredients
- Remain stable for up to 24 hours after activation
Specifications
Specimen Collection Kit | |
CE | |
For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis | |
15°C to 25°C |