missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: CHROMagar™ MRSA II
For the qualitative direct detection of nasal colonization by methicillin-resistant Staphylococcus aureus (MRSA)
$151.42 - $151.42
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Amber |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
22-350-783
|
BD
215228 |
20/Pk. |
N/A
|
|
||||||
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Chromopeptone 35g/L, Chromogen Mix 0.5g/L, Sodium Chloride 17.5g/L, Inhibitory Agents 7.5g/L, Cefoxitin 5.2g/L, Agar 14g/L | |
Plate |
Amber | |
Liquid | |
Chromagar™ MRSA II | |
Selective and differential chromogenic medium for the qualitative direct detection of methicillin resistant Staphylococcus aureus from anterior nares specimens | |
450 g |