missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ BCA Protein Assay Kit

This BCA Protein Assay Kit is used to determine protein concentrations from various sample types.

Supplier:  MP Biomedicals™ 08W00021

Catalog No. ICN08W00021


Explore available promotions

Description

Description

The two-component assay kit can be utilized to evaluate total protein yields in cell lysates, affinity column fractions, purified protein samples, and to monitor contaminants in industrial applications. The BCA protein assay has good compatibility with samples that contain up to 5% surfactants (detergents), such as SDS, Triton X-100, Tween-20, and Tween-80. It is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.

  • High precision protein estimation
  • Detergent-compatible
  • Assay detection sensitivity from 25 to 1000 μg/mL
  • Colorimetric- estimate protein concentrations using a plate reader
  • 20 to 30 minutes total assay time
Specifications

Specifications

Room Temperature
Colorimetric
Assay Kit
25 to 1000 μg/mL
1 Kit
Protein Assay Kit
Colorimetric Detection Kit
Protein
2500 Samples
Protein
Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.