BD BBL™ Cefinase™ β-Lactamase Detection Discs
Each disc is used to test a single bacterial strain for the presence of Β-lactamase
Supplier:  BD 231650
Catalog No. B31650
Explore available promotions
Description
Description
Rapid test used to isolate colonies of Neisseria gonorrhoeae, Staphylococcus sp., Haemophilus influenzae and anaerobic bacteria for the production of Β-lactamase:
  • Quarter inch disc impregnated with nitrocefin, a chromogenic cephalosporin
  • When a bacterium produces Β-lactamase in significant quantities, the yellow-colored disc turns red in the area where the isolate is smeared
Specifications
Specifications
Nitrocefin
-20°C to +8°C
Disc
50 Discs