BD BBL™/Difco™ TB Brilliant Green K
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Supplier: BD 212523
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
TB Brilliant Green K | |
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures | |
Bottle |
4 x 250 mL | |
15° to 30°C | |
Brilliant Green K |
For in vitro Diagnostic Use.