missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Mueller Hinton II Broth (Cation-Adjusted) Prepared Media
For use in quantitative procedures for susceptibility testing of aerobic and facultatively anaerobic bacteria from clinical specimens
$699.03 - $699.03
Specifications
| Certifications/Compliance | ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9 |
|---|---|
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Mueller Hinton II Broth |
| Formulation | Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L |
Description
- Formulated to have low thymine and thymidine content
- Consisting of microorganisms, nutrient medium and antimicrobial agent
Specifications
| ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9 | |
| United States | |
| Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L |
| Liquid | |
| Mueller Hinton II Broth |
For in vitro Diagnostic Use.