missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated: CHROMagar™ Orientation Media
Nonselective, differential medium for presumptively identifying bacterial isolates from primary clinical specimens
$194.63 - $194.63
Specifications
| Form | Liquid |
|---|---|
| Manufacturing Origin | United States |
| Formulation | Chromopeptone 16.1g/L, Chromogen Mix 1.3g/L, Agar 15g/L |
| For Use With (Application) | Isolation, differentiation and enumeration of urinary tract pathogens |
| Packaging Type | Plate |
| Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | Color | Media Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | Color | Media Type | Price | Quantity | ||
|
B215081
|
BD
215081 |
100/Pk. | ISO/DIS, 11014 | Light Beige | Orientation Agar |
Pack of 100 Each for $194.63
|
|
||
Description
- Visual differentiationn of colonies is enhanced
- Allows isolation of both Gram positive and Gram negative pathogens
Specifications
| Liquid | |
| Chromopeptone 16.1g/L, Chromogen Mix 1.3g/L, Agar 15g/L | |
| Plate |
| United States | |
| Isolation, differentiation and enumeration of urinary tract pathogens |