missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: BCYE Selective Agar
For isolation of Legionella species from clinical and nonclinical specimens
Supplier: BD 297879
Specifications
| ISO/DIS, 11014 | |
| 2°C to 8°C | |
| United States | |
| BCYE Agar 38.65g/L, Polymyxin B 40000U, Anisomycin 80mg/L, Cefamandole 4mg/L | |
| Plate |
| Dark Green | |
| Liquid | |
| BCYE selective Agar with PAC | |
| Legionella species from contaminated clinical specimens (with cefamandole) | |
| 10/Pk. |