missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: Campy CVA Agar
Selective media used in the primary isolation and cultivation of Campylobacter jejuni from human fecal specimens
Supplier: BD 297246
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Liquid | |
Campylobacter Cefoperazone Vancomycin-Amphotericin B Agar | |
Campylobacter jejuni from Human feces | |
20/Pk. |
Dark Red | |
United States | |
Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Cefoperazone 20mg/L, Vancomycin 10mg/L, Amphotericin B 2mg/L, Sheep Blood, defibrinated | |
Plate | |
1130 g |