missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: Corn Meal Agar with Polysorbate 80 (Deep Fill)
For the testing of Candida species for their ability to produce chlamydospores and for the cultivation of fungi
Supplier: BD 221854
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
- Deep-filled plates reduce effects of drying during prolonged incubation
Specifications
ISO/DIS, 11014 | |
Liquid | |
Corn Meal Agar with Polysorbate 80 | |
Testing Candida species producing Chlamydospores and general purpose for cultivating fungi | |
10/Pk. |
Light Beige | |
United States | |
Corn Meal infusion from (solids) 2g/L, Agar 15g/L, Polysorbate 80 10mL | |
Plate | |
1300 g |