BD BBL™ Prepared Plated Media: MacConkey II Agar
Selective and differential media for the detection of coliform organisms and enteric pathogens
Supplier:  BD 221270
 View more versions of this product
Catalog No. B21270
Explore available promotions
Description
Description
  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
ISO/DIS, 11014
2°C to 8°C
United States
Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L
Plate
Light Beige
Solid
MacConkey II Agar
Coliform organisms and enteric pathogens
100/Pk.