missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Replacement Reagents for Gram Stain Kits

For BBL Gram Stain Kits

$345.03 - $1422.50

Specifications

Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Storage Requirements 15° to 30°C
View More Specs

Products 6
Catalog Number Mfr. No. Type Quantity Stain Type Price Quantity  
Catalog Number Mfr. No. Type Quantity Stain Type Price Quantity  
B4312531
View Documents
BD
212531
Safranin 4 x 250mL Safranin
Case of 4 Each for $345.03
 
B4312532
View Documents
BD
212532
Safranin 3.8L Safranin
Each for $539.37
 
B4312542
View Documents
BD
212542
Gram Iodine 4 x 250mL Iodine
Case of 4 Each for $353.47
 
B4312543
View Documents
BD
212543
Gram Iodine (Stabilized) 3.8L Iodine
Each for $741.29
 
B4312544
View Documents
BD
212544
Basic Fuchsin 4 x 250mL Basic Fuchsin
Pack of 4 Each for $616.97
 
B4312545
View Documents
BD
212545
Basic Fuchsin 3.8L Basic Fuchsin
Each for $1,422.50
 
Description

Description

  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
  • Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles; sales of B12542 are restricted from California

Specifications

Specifications

CE
Used to stain microorganisms from cultures or specimens by the differential gram method
15° to 30°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'