BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Novobiocin (30μg)
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Supplier:  BD 231315
Catalog No. B31315
Explore available promotions
Description
Description
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Novobiocin
30μg
Disc
10 Pack
Videos
SDS
Documents
Documents
Accessories