BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Penicillin (10U)
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
$23.92 - $138.82
Specifications
Antimicrobial Agents Penicillin
Concentration 10U
Packaging Type Disc
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30918
View Documents
BD
230918
1/Pk.
Each for $23.92
B31321 Edge
Accessories View Documents
BD
231321
10/Pk.
Case of 10 Each for $138.82
Edge
Description
Description
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
Specifications
Specifications
Penicillin
10U
Disc
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'