missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Catheter-Tip Syringes
Bulk, nonsterile syringes with catheter tip
Supplier: BD 301037
Alert:
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Description
- Volume: 50 mL
- Shelf-life: 1825 days
Specifications
| Catheter-tip Syringe | |
| 50 mL | |
| Non-sterile |