missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Baird-Parker Agar Base
Use with EY Tellurite Enrichment for the detection and enumeration of coagulase positive staphylococci in foods and other materials
Supplier: BD 276840
Description
- Formulation: Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Lithium Chloride 5g/L, Agar 20g/L
Specifications
| <30°C | |
| Baird-Parker Agar Base | |
| Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Lithium Chloride 5g/L, Agar 20g/L | |
| Bottle |
| Dehydrated | |
| Agar | |
| For coagulase positive Staphylococcus | |
| 500 g |