missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment
For injured Salmonella in food
Supplier: BD 218105
Specifications
| ISO/DIS, 11014, BAM, CCAM, ISO, USDA | |
| Dehydrated | |
| Peptone Water | |
| Isolation of Salmonella species | |
| 500 g |
| <30°C | |
| Peptone Water | |
| Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L | |
| Bottle |