missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Demi-Fraser Broth Base
For use with Fraser broth supplement in selectively and differentially enriching Listeria, reduced Acriflavine and Nalidixic Acid levels
Supplier: BD 265310
Specifications
| 2°C to 8°C | |
| Demi-Fraser Broth Base | |
| Tryptose 10g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.01g/L, Acriflavine HCl 12.5mg/L, Lithium Chloride 3g/L | |
| Pail |
| Dehydrated | |
| Microbiology Media | |
| For use with Fraser broth supplement in selectively and differentially enriching Listeria, reduced Acriflavine and Nalidixic Acid levels | |
| 10,000 g |