missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani)

Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Supplier:  BD B244510

 View more versions of this product

Catalog No. DF0445-07-6


Explore available promotions

Description

Description

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications

 

Specifications

Specifications

CCAM
Dehydrated
Agar
Propagation and maintenance of Escherichia coli for molecular biology
2000 g
<30°C
LB Agar, Miller
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L
Pail
SDS
Documents

Documents

Promotions

Promotions