missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: mFC Broth Base
For cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature
$455.42 - $455.42
Specifications
Certifications/Compliance | CE, EPA, SMWW |
---|---|
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | mFC Broth Base |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
DF0883-17-3
|
BD
288330 |
500 g |
Each for $455.42
|
|
||||||
Description
- Use with Rosolic Acid
- Formulation: Tryptose 10g/L, Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Aniline blue 0.1g/L
Certifications
CE, EPA, SMWW
Specifications
CE, EPA, SMWW | |
Dehydrated | |
Microbiology Media | |
Used with Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature |
Refrigerate | |
mFC Broth Base | |
Tryptose 10g/L, Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Aniline blue 0.1g/L | |
Bottle |