BD Difco™ Salmonella H Antiserum z
Qualitative tube agglutination tests for serological differentiation of Salmonella
Supplier: BD 222771
Alert:
The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.
Description
- Lyophilized and polyclonal rabbit antisera
- Absorbed or unabsorbed specifically for either phase 1 or phase 2 antigens
- Perform between 150-1500 tests
- Identification of Salmonella by flagellar (H) antigens
- Sodium azide (0.2%) used as preservative
Specifications
Antiserum | |
3 mL |