BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
$51.66 - $115.00
Specifications
Product Type Syringe
Disposable Single-use
Connection Type Luer Lock
Sterility Sterile
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
14-823-30
View Documents
BD
309628
1 mL
Pack of 100 Each for $115.00
Case of 8 Pack for $920.00
B309646 Edge
View Documents
BD
309646
5 mL
Pack of 125 Each for $59.09
Case of 4 Pack for $236.36
Edge
B309657 Edge
View Documents
BD
309657
3 mL
Pack of 200 Each for $51.66
Case of 4 Pack for $206.64
Edge
Description
Description
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
Specifications
Specifications
Syringe
Single-use
Luer Lock
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'