BD GasPak™ EZ Gas Generating Systems Incubation Containers
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Supplier:  BD 260002
 View more versions of this product
Catalog No. B260002
Explore available promotions
Description
Description
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
  • Non-breakable, chemical resistant containers
  • Removable Petri dish racks are available (sold separately)
  •  
Specifications
Specifications
EZ Incubation Container
2°C to 25°C
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
GasPak
Easy close latches
Reusable
Petri Dishes (10 to 12)
Small