BD Macro-Vue™ Model 51 RPR Card Test Rotator Package
For use with BD Macro-Vue RPR and other card test kits
Supplier:  BD 278051
Catalog No. B4978051
Explore available promotions
Description
Description
  • Rotator Package L4978051 contains one Rotator, a 5 x 7 in. Platform, and one Humidifying Cover
  • Also available is an Accessory Package L4977979 that contains one 7 x 15 in rotator platform and two humidifying covers
Specifications
Specifications
100 rpm
Rotator Package
1 Humidifying cover
Product Suggestions
Product Suggestions